Learn More
AIMS™ Hypothermic Cooling Plate
SCI_ED
Catalog No. 14370224
Change view
Click to view available options
For Use With (Equipment):
Grace Small Animal Surgery Boards
Catalog No. For Use With (Equipment)
14-370-224 Grace Small Animal Surgery Boards
1 options
Catalog No. 14-370-224 Supplier AIMS™ Supplier No. GBHP
Add to Cart
Add to Cart
Use with ice to cool the plate to 3anddeg; to 4anddeg;C
  • Equipped with cooling fins on the bottom
  • For Grace Small Animal Surgery Boards
Specifications
For Use With (Application) Use with ice to cool the plate to 3° to 4°C
Type Hypothermic Cooling Plate
For Use With (Equipment) Grace Small Animal Surgery Boards
Includes Cooling fins on the bottom