Learn More
BIOLIFE SOLUTIONS LLC HypoThermosol® 10mL
SDP
Catalog No. NC1067910
Encompass
This item has a minimum qty of 6 per supplier requirements.
HypoThermosol is an optimized hypothermic (2-8C) preservation media that enables improved and extended preservation of cells, tissues and organs. HypoThermosol is uniquely formulated to address the molecular-biological response of cells during the hypothermic preservation process. HypoThermosol includes key ions at concentrations that balance the intracellular state at hypothermic temperatures. Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers.
Catalog No. NC1067910 Supplier BIOLIFE SOLUTIONS LLC Supplier No. 101373
This item has a minimum qty of 6 per supplier requirements.
HypoThermosol is an optimized hypothermic (2-8C) preservation media that enables improved and extended preservation of cells, tissues and organs. HypoThermosol is uniquely formulated to address the molecular-biological response of cells during the hypothermic preservation process. HypoThermosol includes key ions at concentrations that balance the intracellular state at hypothermic temperatures. Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers.