Learn More
STEMCELL Technologies HypoThermosol FRS, Size: 500 mL
SDP
Supplier:  STEMCELL Technologies 07945
Encompass
Improve and extend the preservation of cells tissues and organs with HypoThermosol FRS. This ready-to-use serum-free protein-free and animal component-free preservation medium is formulated to address the molecular-biological response of cells during the hypothermic (2 - 8C) preservation process. HypoThermosol FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures. This medium has been subjected to sterility endotoxin and cell-based release testing and is manufactured under cGMPs with USP-grade components. Additional components include pH buffers energy substrates free radical scavengers and osmotic/oncotic stabilizers. Visit STEMCELL.com for more information.
Catalog No. 50-239-0311