Learn More
Charles River Labs Cell Solutions HypoThermosol FRS Biopreservation Media, 500mL Bottle
Catalog No. 502256130
Encompass
HypoThermosol is a uniquely formulated serum-free and animal component-free biopreservation medium that contains key ions, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers, thus enabling improved and extended preservation of cells at hypothermic temperatures ranging between 2-8°C. HypoThermosol is cGMP manufactured with USP grade components.
This item is not returnable. View return policy
Catalog No. 50-225-6130 Supplier Charles River Labs Cell Solutions Supplier No. BPM 101104
HypoThermosol is a uniquely formulated serum-free and animal component-free biopreservation medium that contains key ions, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers, thus enabling improved and extended preservation of cells at hypothermic temperatures ranging between 2-8°C. HypoThermosol is cGMP manufactured with USP grade components.