Learn More
STEMCELL Technologies HypoThermosol FRS, Size: 160 mL
SDP
Catalog No. 501974434
Encompass
Improve and extend the preservation of cells, tissues, and organs with HypoThermosol FRS. This ready-to-use, serum-free, protein-free, and animal component-free preservation medium is formulated to address the molecular-biological response of cells during the hypothermic (2 - 8°C) preservation process. HypoThermosol FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures. This medium has been subjected to sterility, endotoxin, and cell-based release testing and is manufactured under cGMPs with USP-grade components. Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers. Visit STEMCELL.com for more information.
Catalog No. 50-197-4434 Supplier STEMCELL Technologies Supplier No. 07934
Improve and extend the preservation of cells, tissues, and organs with HypoThermosol FRS. This ready-to-use, serum-free, protein-free, and animal component-free preservation medium is formulated to address the molecular-biological response of cells during the hypothermic (2 - 8°C) preservation process. HypoThermosol FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures. This medium has been subjected to sterility, endotoxin, and cell-based release testing and is manufactured under cGMPs with USP-grade components. Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers. Visit STEMCELL.com for more information.