Learn More
Cayman Chemical Hypoxanthine, Purity: ≥98%, CAS: 68-94-0, 25 g
SDP
Catalog No. 502986987
Encompass_Preferred
An endogenous purine derivative; intrastriatal administration increases mitochondrial complex II, also known as succinate dehydrogenase, activity and decreases cytochrome c oxidase activity, resulting in neuroenergetic impairment, ATP depletion, and cellular apoptosis in rats at 10 µM; induces hyperuricemia in mice; spinal fluid levels are increased in patients with Lesh-Nyhan syndrome
Catalog No. 50-298-6987 Supplier Cayman Chemical Supplier No. 2225425 G
An endogenous purine derivative; intrastriatal administration increases mitochondrial complex II, also known as succinate dehydrogenase, activity and decreases cytochrome c oxidase activity, resulting in neuroenergetic impairment, ATP depletion, and cellular apoptosis in rats at 10 µM; induces hyperuricemia in mice; spinal fluid levels are increased in patients with Lesh-Nyhan syndrome