Learn More
Selleck Chemical LLC Hypoxanthine S2424-25mg
SDP
Catalog No. 503108830
Encompass_Preferred
Hypoxanthine a naturally occurring purine derivative an intermediate product of uric acid synthesis formed from adenylic acid and itself a precursor of xanthine
Catalog No. 50-310-8830 Supplier Selleck Chemical LLC Supplier No. S242425MG
Hypoxanthine a naturally occurring purine derivative an intermediate product of uric acid synthesis formed from adenylic acid and itself a precursor of xanthine