Learn More
Medchemexpress LLC HY-N0091 50mg Medchemexpress, Hypoxanthine CAS:68-94-0 Purity:>98%
SDP
Catalog No. 501933385
Encompass_Preferred
Medchemexpress, HY-N0091 50mg Hypoxanthine CAS:68-94-0 Hypoxanthine, a purine derivative, is a potential free radical generator and could be used as an indicator of hypoxia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3385 Supplier Medchemexpress LLC Supplier No. HYN009150MG
Medchemexpress, HY-N0091 50mg Hypoxanthine CAS:68-94-0 Hypoxanthine, a purine derivative, is a potential free radical generator and could be used as an indicator of hypoxia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.