Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hypoxanthine-d4 | 2483831-32-7 | 99.7% | 140.14 | C5D4N4O | 10 MG
SDP

Catalog No. 5000295973
Encompass_Preferred

Hypoxanthine-d4 is the deuterium-labeled analog of hypoxanthine used as an analytical internal standard and tracer in mass spectrometry and biochemical assays. It provides a stable isotope reference for quantification in studies of purine metabolism and hypoxia.

  • Deuterium-labeled internal standard for LC/GC-MS quantification.
  • High purity (99.7%) suitable for analytical applications.
  • Molecular formula C5D4N4O and molecular weight 140.14 g/mol.
  • Soluble in DMSO at 10 mg/mL; ultrasonic and warming recommended.
  • Store sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-95973 Supplier Medchemexpress LLC Supplier No. HYN0091S510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hypoxanthine-d4 is the deuterium-labeled analog of hypoxanthine used as an analytical internal standard and tracer in mass spectrometry and biochemical assays. It provides a stable isotope reference for quantification in studies of purine metabolism and hypoxia.

  • Deuterium-labeled internal standard for LC/GC-MS quantification.
  • High purity (99.7%) suitable for analytical applications.
  • Molecular formula C5D4N4O and molecular weight 140.14 g/mol.
  • Soluble in DMSO at 10 mg/mL; ultrasonic and warming recommended.
  • Store sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.