Learn More
EUROTROL HYPOXIC QC XLOW, 5 AMPULES/BOX
Catalog No. NC0882958
Encompass
CueSee Hypoxic. Clear ampules with One Point Cut containing 2.5 mL purified stroma-free bovine hemoglobin. Shelf life is 1 month when stored at 2 - 8 deg.C, calculated from shipping date to customer.
Catalog No. NC0882958 Supplier EUROTROL Supplier No. 166.001.005
CueSee Hypoxic. Clear ampules with One Point Cut containing 2.5 mL purified stroma-free bovine hemoglobin. Shelf life is 1 month when stored at 2 - 8 deg.C, calculated from shipping date to customer.