Learn More
Cayman Chemical Hypoxanthine-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2483831-32-7, 50 mg
SDP
Catalog No. 502912084
Encompass_Preferred
An internal standard for the quantification of hypoxanthine by GC- or LC-MS
Catalog No. 50-291-2084 Supplier Cayman Chemical Supplier No. 3633050 MG
An internal standard for the quantification of hypoxanthine by GC- or LC-MS