Learn More
Medchemexpress LLC HY-A0104 1g Medchemexpress, Hypromellose Celacol HPM 5000;(Hydroxypropyl)methyl cellulose;HPMC CAS:9004-65-3 Purity:98%
SDP
Catalog No. 501639116
Encompass_Preferred
Medchemexpress,HY-A0104 1g Hypromellose Celacol HPM 5000;(Hydroxypropyl)methyl cellulose;HPMC CAS:9004-65-3 Formula:N/A Hypromellose is a water-soluble hydrophilic, non-ionic cellulose ether used to form swellable-soluble matrices. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9116 Supplier Medchemexpress LLC Supplier No. HYA01041G
Medchemexpress,HY-A0104 1g Hypromellose Celacol HPM 5000;(Hydroxypropyl)methyl cellulose;HPMC CAS:9004-65-3 Formula:N/A Hypromellose is a water-soluble hydrophilic, non-ionic cellulose ether used to form swellable-soluble matrices. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.