Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hyptadienic acid | 128397-09-1 | MFCD17214838 | 93.0% | 470.7 g/mol | C30H46O4 | 10 MG
SDP

Catalog No. 5000289768
Encompass_Preferred

Hyptadienic acid is a triterpene carboxylic acid isolated from Perilla frutescens, provided as a solid reagent for research use in biochemical and pharmacological studies of inflammation.

  • Triterpene acid used for research of inflammation.
  • Inhibits TPA-induced inflammation in animal models.
  • Purity approximately 93.0% suitable for research applications.
  • Molecular weight around 470.7 g/mol, molecular formula C30H46O4.
  • Appearance: solid, white to off-white.
  • Available in small research sizes such as 10 mg.
  • Store protected from light; short-term storage at -20°C, long-term in solvent at -80°C.

Catalog No. 50-002-89768 Supplier Medchemexpress LLC Supplier No. HYN402410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hyptadienic acid is a triterpene carboxylic acid isolated from Perilla frutescens, provided as a solid reagent for research use in biochemical and pharmacological studies of inflammation.

  • Triterpene acid used for research of inflammation.
  • Inhibits TPA-induced inflammation in animal models.
  • Purity approximately 93.0% suitable for research applications.
  • Molecular weight around 470.7 g/mol, molecular formula C30H46O4.
  • Appearance: solid, white to off-white.
  • Available in small research sizes such as 10 mg.
  • Store protected from light; short-term storage at -20°C, long-term in solvent at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.