Learn More
MSC Loctite 237099 50 mL Cartridge Two Part Epoxy 3 min Working Time, 1,430 psi Shear Strength, Series E-05CL
Catalog No. 501149326
Encompass
Loctite 237099 50 mL Cartridge Two Part Epoxy 3 min Working Time, 1,430 psi Shear Strength, Series E-05CL
Catalog No. 50-114-9326 Supplier MSC Supplier No. 00611574
Loctite 237099 50 mL Cartridge Two Part Epoxy 3 min Working Time, 1,430 psi Shear Strength, Series E-05CL