Learn More
Advanced Biomatrix HyStem Version 2.0 Thiol-Modified Hyaluronan Hydrogel Kit, 12.5 mL
Catalog No. NC0897704
Encompass_Preferred
HyStem contains thiolated hyaluronic acid for easy 3D cell culture. HyStem version 2.0 is reconstituted with a buffered 1X PBS. The resulting components will also result in a pH neutral isotonic salt hydrogel. The ending pH, osmo, and rheological properties are the same from Version 1.0 to Version 2.0. HyStem hydrogels are xeno-free and completely customizable. The hydrogel is formed when the crosslinking agent, Extralink-Lite (PEGDA) is added to a mixture of Glycosil (thiol-modified hyaluronan). Recommended for applications requiring specific attachment factor optimization as well as non-adherent cell culture, cellular applications. Extracellular matrix proteins can be mixed into the hydrogel and incorporated non-covalently before gelation. The HyStem Hydrogel Kits are optimal for culturing stem cells whose natural environment is rich in hyaluronic acid (HA).
Catalog No. NC0897704 Supplier Advanced Biomatrix Supplier No. GS1004F
HyStem contains thiolated hyaluronic acid for easy 3D cell culture. HyStem version 2.0 is reconstituted with a buffered 1X PBS. The resulting components will also result in a pH neutral isotonic salt hydrogel. The ending pH, osmo, and rheological properties are the same from Version 1.0 to Version 2.0. HyStem hydrogels are xeno-free and completely customizable. The hydrogel is formed when the crosslinking agent, Extralink-Lite (PEGDA) is added to a mixture of Glycosil (thiol-modified hyaluronan). Recommended for applications requiring specific attachment factor optimization as well as non-adherent cell culture, cellular applications. Extracellular matrix proteins can be mixed into the hydrogel and incorporated non-covalently before gelation. The HyStem Hydrogel Kits are optimal for culturing stem cells whose natural environment is rich in hyaluronic acid (HA).