Learn More
Hyvac Products Inc HYVAC FLUSHING OIL - 1QT
Catalog No. 50465192
Encompass_Preferred
Oil, Flushing; Hyvac; Safely removes sediment, contamination from high vacuum pumps; One or two flushings cleans residue, protects pump; Pump can be refilled immediately after flushing; Should not be used for high vacuum operation; 1 Quart.
Catalog No. 50-465-192 Supplier Hyvac Products Inc Supplier No. 93060001
Oil, Flushing; Hyvac; Safely removes sediment, contamination from high vacuum pumps; One or two flushings cleans residue, protects pump; Pump can be refilled immediately after flushing; Should not be used for high vacuum operation; 1 Quart.