Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coast HZ050 Safety-Rated Flashlight

Catalog No. 19012087
Change view
Click to view available options
Color:
Hi-vis Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-012-087 Hi-vis Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-012-087 Supplier Coast Supplier No. 30047

missing translation for 'validMainframeMsg'

Flashlight combines LED beam-shaping technology with five different beam options for maximum visibility in Class 1, Division 1 environments.

  • Intrinsically Safe light shines bright light even in the most hazardous work environments
  • Combines LED beam-shaping technology with Intrinsically Safe certification (Class I, Division 1)
  • Ideal source of high-powered light even under the toughest, riskiest conditions
  • Features 5 beam options including Spot and Flood beams

Specifications

Weight (English) 8.1 oz.
Weight (Metric) 230 g
Battery Type 3 x AA (Alkaline)
Certifications/Compliance Class I, II & III, Division 1, GP A-G T3 CL I, ZN 0, Ex&AEx ia IIC T3 Ga
Color Hi-vis Yellow
Description Safety-Rated
Diameter (English) Body 1.81 in.
Diameter (Metric) Body 4.6 cm
Includes 3 x AA Batteries, Battery Key, and Magnet Clip
Length (English) 7.5 in.
Length (Metric) 19.1 cm
Product Type Flashlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.