Learn More
Medchemexpress LLC HZX-02-059 | 2240205-30-3 | 99.4% | 487.48 | 100 MG
SDP
Catalog No. 5000438453
Encompass_Preferred
HZX-02-059 is a potent methuosis inducer and a dual-target PIKfyve/tubulin inhibitor with anticancer activity. It primarily disrupts the balance of endocytosis and exocytosis, leading to the formation of numerous vesicles and inducing cell death. Additionally, HZX-02-059 promotes cell vacuolization, apoptosis, downregulates the p53 pathway, inhibits PI3K/AKT phosphorylation, and suppresses c-Myc and NF-κB transcription.
  • Potent methuosis inducer.
  • Dual-target PIKfyve/tubulin inhibitor.
  • Exhibits anticancer activity.
  • Induces cell vacuolization.
  • Promotes apoptosis.
  • Downregulates the p53 pathway.
  • Inhibits PI3K/AKT phosphorylation.
  • Inhibits c-Myc and NF-κB transcription.
Catalog No. 50-004-38453 Supplier Medchemexpress LLC Supplier No. HY112440100MG
HZX-02-059 is a potent methuosis inducer and a dual-target PIKfyve/tubulin inhibitor with anticancer activity. It primarily disrupts the balance of endocytosis and exocytosis, leading to the formation of numerous vesicles and inducing cell death. Additionally, HZX-02-059 promotes cell vacuolization, apoptosis, downregulates the p53 pathway, inhibits PI3K/AKT phosphorylation, and suppresses c-Myc and NF-κB transcription.
  • Potent methuosis inducer.
  • Dual-target PIKfyve/tubulin inhibitor.
  • Exhibits anticancer activity.
  • Induces cell vacuolization.
  • Promotes apoptosis.
  • Downregulates the p53 pathway.
  • Inhibits PI3K/AKT phosphorylation.
  • Inhibits c-Myc and NF-κB transcription.