Learn More
Medchemexpress LLC HZX-02-059 | 2240205-30-3 | 99.4% | 487.48 | 50 MG
SDP
Catalog No. 5000436878
Encompass_Preferred
HZX-02-059 is a potent methuosis inducer and dual-target PIKfyve/tubulin inhibitor with anticancer activity. Methuosis primarily disrupts the balance of endocytosis and exocytosis, leading to the formation of numerous vesicles and inducing cell death. This compound also induces cell vacuolization, promotes apoptosis, downregulates the p53 pathway, inhibits PI3K/AKT phosphorylation, and inhibits c-Myc and NF-κB transcription.
  • Potent methuosis inducer
  • Dual-target PIKfyve/tubulin inhibitor
  • Anticancer activity
  • Induces cell vacuolization
  • Promotes apoptosis
  • Downregulates the p53 pathway
  • Inhibits PI3K/AKT phosphorylation
  • Inhibits c-Myc and NF-κB transcription
Catalog No. 50-004-36878 Supplier Medchemexpress LLC Supplier No. HY11244050MG
HZX-02-059 is a potent methuosis inducer and dual-target PIKfyve/tubulin inhibitor with anticancer activity. Methuosis primarily disrupts the balance of endocytosis and exocytosis, leading to the formation of numerous vesicles and inducing cell death. This compound also induces cell vacuolization, promotes apoptosis, downregulates the p53 pathway, inhibits PI3K/AKT phosphorylation, and inhibits c-Myc and NF-κB transcription.
  • Potent methuosis inducer
  • Dual-target PIKfyve/tubulin inhibitor
  • Anticancer activity
  • Induces cell vacuolization
  • Promotes apoptosis
  • Downregulates the p53 pathway
  • Inhibits PI3K/AKT phosphorylation
  • Inhibits c-Myc and NF-κB transcription