Learn More
Medchemexpress LLC HZX-02-059 (methuosis inducer 1) | 2240205-30-3 | 99.4% | 487.48 | 10 MG
SDP
Catalog No. 5000437408
Encompass_Preferred
HZX-02-059 is a potent methuosis inducer and a dual-target PIKfyve/tubulin inhibitor, demonstrating anticancer activity. This compound primarily disrupts the balance of endocytosis and exocytosis, leading to the formation of numerous vesicles and inducing cell death.
  • Induces cell vacuolization and promotes apoptosis
  • Downregulates the p53 pathway
  • Inhibits PI3K/AKT phosphorylation
  • Inhibits c-Myc and NF-κB transcription
  • Exhibits broad spectrum cytotoxicity against various human cancer cell lines
  • For research use only
Catalog No. 50-004-37408 Supplier Medchemexpress LLC Supplier No. HY11244010MG
HZX-02-059 is a potent methuosis inducer and a dual-target PIKfyve/tubulin inhibitor, demonstrating anticancer activity. This compound primarily disrupts the balance of endocytosis and exocytosis, leading to the formation of numerous vesicles and inducing cell death.
  • Induces cell vacuolization and promotes apoptosis
  • Downregulates the p53 pathway
  • Inhibits PI3K/AKT phosphorylation
  • Inhibits c-Myc and NF-κB transcription
  • Exhibits broad spectrum cytotoxicity against various human cancer cell lines
  • For research use only