Learn More
BUCHI I-100 Interface
SDP
Catalog No. 05405192 Shop All BUCHI Products
Change view
Click to view available options
Includes:
Power Pack, Communication Cable (Mini-DIN), and Holder to Fasten to Lab-Stand
Power Pack, Valve Unit, and Holder to Fasten to Lab-Stand
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
05-405-192 Power Pack, Valve Unit, and Holder to Fasten to Lab-Stand
05-405-122 Power Pack, Communication Cable (Mini-DIN), and Holder to Fasten to Lab-Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-405-192 Supplier BUCHI Supplier No. 11I100003
The I-100 is an easy to use interface to conveniently regulate vacuum. As a central interface of the Rotavapor R-100, it increases the efficiency of the evaporation process.
  • Functional design fulfills essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to centralized control of major process parameters
  • Full process control thanks to parameter monitoring
  • Proven technology for major applications
Specifications
Certifications/Compliance CE
Material PBT
Type Vacuum Controller
For Use With (Equipment) Vacuum Pump Model V-100
Includes Power Pack, Valve Unit, and Holder to Fasten to Lab-Stand
Model I-100
Length (Metric) 160 mm
Voltage 24 V