Learn More
Medchemexpress LLC I-138 | 2098211-50-6 | C26H23F3N6O | 100 MG
SDP
Catalog No. 5000372819
Encompass_Preferred
I-138 is an orally active, reversible inhibitor of USP1-UAF1. It induces monoubiquitination of FANCD2 and PCNA in cells and eliminates USP1 autocleavage. This compound is structurally related to ML323.
  • Orally active and reversible inhibitor of USP1-UAF1
  • Induces monoubiquitination of FANCD2 and PCNA in cells
  • Eliminates USP1 autocleavage in cells
  • Shows dose-dependent inhibition of MDA-MB-436 cell viability
  • Exhibits modest antitumor activity in mice bearing MDA-MB-436 tumors
  • Can be combined with Niraparib for BRCA1/2 mutant tumor inhibition in vivo
Catalog No. 50-003-72819 Supplier Medchemexpress LLC Supplier No. HY153365100MG
I-138 is an orally active, reversible inhibitor of USP1-UAF1. It induces monoubiquitination of FANCD2 and PCNA in cells and eliminates USP1 autocleavage. This compound is structurally related to ML323.
  • Orally active and reversible inhibitor of USP1-UAF1
  • Induces monoubiquitination of FANCD2 and PCNA in cells
  • Eliminates USP1 autocleavage in cells
  • Shows dose-dependent inhibition of MDA-MB-436 cell viability
  • Exhibits modest antitumor activity in mice bearing MDA-MB-436 tumors
  • Can be combined with Niraparib for BRCA1/2 mutant tumor inhibition in vivo