Learn More
Selleck Chemical LLC I-138-E1479-5MG
SDP
Catalog No. 503136933
Encompass_Preferred
I-138 is an orally active potent reversible inhibitor of USP1-UAF1 with IC50 0f 4 1 nM I-138 is structurally related to ML323 and induces the monoubiquitination of FANCD2 and PCNA in MDA-MB-436 cells and increases PCNA and FANCD2 monoubiquitination in HAP-1 USP1 WT cells
Catalog No. 50-313-6933 Supplier Selleck Chemical LLC Supplier No. E14795MG
I-138 is an orally active potent reversible inhibitor of USP1-UAF1 with IC50 0f 4 1 nM I-138 is structurally related to ML323 and induces the monoubiquitination of FANCD2 and PCNA in MDA-MB-436 cells and increases PCNA and FANCD2 monoubiquitination in HAP-1 USP1 WT cells