Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC I-191 | 1690172-25-8 | 99.1% | 423.48 | C23H26FN5O2 | 1 ML
SDP

Catalog No. 5000227599
Encompass_Preferred

I-191 is a potent, selective antagonist of protease-activated receptor 2 (PAR2) for research use. It attenuates PAR2-mediated intracellular signaling, including Ca2+ release and ERK1/2 phosphorylation, and is supplied as a high-purity solid or a ready-to-use DMSO solution.

  • Molecular weight: 423.48 g/mol.
  • Chemical formula: C23H26FN5O2.
  • CAS number: 1690172-25-8.
  • Purity: 99.14%.
  • Form and color: solid; white to gray.
  • Solubility: DMSO 8.33 mg/mL (19.67 mM); ultrasonic agitation recommended.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).
  • Supplied quantities: 10 mM, 1 mL solution; solids 5 mg-100 mg (larger sizes available).

Catalog No. 50-002-27599 Supplier Medchemexpress LLC Supplier No. HY11779310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

I-191 is a potent, selective antagonist of protease-activated receptor 2 (PAR2) for research use. It attenuates PAR2-mediated intracellular signaling, including Ca2+ release and ERK1/2 phosphorylation, and is supplied as a high-purity solid or a ready-to-use DMSO solution.

  • Molecular weight: 423.48 g/mol.
  • Chemical formula: C23H26FN5O2.
  • CAS number: 1690172-25-8.
  • Purity: 99.14%.
  • Form and color: solid; white to gray.
  • Solubility: DMSO 8.33 mg/mL (19.67 mM); ultrasonic agitation recommended.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).
  • Supplied quantities: 10 mM, 1 mL solution; solids 5 mg-100 mg (larger sizes available).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.