Learn More
Medchemexpress LLC I-191 | 1690172-25-8 | 99.1% | 423.48 | C23H26FN5O2 | 50 MG
SDP
Catalog No. 5000226545
Encompass_Preferred
I-191 is a potent, selective protease-activated receptor 2 (PAR2) antagonist used as a research reagent to block PAR2-mediated signaling pathways in cell-based assays, resulting in reduced cytokine production and downstream effects such as Ca2+ mobilization and ERK1/2 phosphorylation.
  • Potent, selective PAR2 antagonist.
  • Inhibits PAR2-mediated Ca2+ release and ERK1/2 phosphorylation.
  • Reduces cytokine production in cell-based assays.
  • High purity (reported ~99.14%).
  • Available in multiple pack sizes, including 50 MG.
Catalog No. 50-002-26545 Supplier Medchemexpress LLC Supplier No. HY11779350MG
I-191 is a potent, selective protease-activated receptor 2 (PAR2) antagonist used as a research reagent to block PAR2-mediated signaling pathways in cell-based assays, resulting in reduced cytokine production and downstream effects such as Ca2+ mobilization and ERK1/2 phosphorylation.
  • Potent, selective PAR2 antagonist.
  • Inhibits PAR2-mediated Ca2+ release and ERK1/2 phosphorylation.
  • Reduces cytokine production in cell-based assays.
  • High purity (reported ~99.14%).
  • Available in multiple pack sizes, including 50 MG.