Learn More
Medchemexpress LLC I-191 | 1690172-25-8 | 99.1% | 423.48 g·mol⁻1 | C23H26FN5O2 | 100 MG
SDP
Catalog No. 5000227813
Encompass_Preferred
I-191 is a potent, selective small-molecule antagonist of protease-activated receptor 2 (PAR2) developed for research use. It inhibits PAR2-mediated signaling and cytokine responses and is provided as a solid research-grade compound.
  • Potent, selective antagonist of protease-activated receptor 2 (PAR2).
  • Demonstrated activity in cell-based assays inhibiting PAR2 signaling and cytokine production.
  • Supplied as a solid research compound suitable for in vitro pharmacology and signal transduction studies.
  • Molecular weight 423.48 g·mol⁻1 and molecular formula C23H26FN5O2.
  • Certificate of analysis and safety data sheet available to support quality and safe handling.
Catalog No. 50-002-27813 Supplier Medchemexpress LLC Supplier No. HY117793100MG
I-191 is a potent, selective small-molecule antagonist of protease-activated receptor 2 (PAR2) developed for research use. It inhibits PAR2-mediated signaling and cytokine responses and is provided as a solid research-grade compound.
  • Potent, selective antagonist of protease-activated receptor 2 (PAR2).
  • Demonstrated activity in cell-based assays inhibiting PAR2 signaling and cytokine production.
  • Supplied as a solid research compound suitable for in vitro pharmacology and signal transduction studies.
  • Molecular weight 423.48 g·mol⁻1 and molecular formula C23H26FN5O2.
  • Certificate of analysis and safety data sheet available to support quality and safe handling.