Learn More
TARGETMOL CHEMICALS INC I-191 2MG
SDP
Catalog No. 502879706
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. I-191 is a potent antagonist of protease-activated receptor 2 (PAR2), inhibits multiple signaling functions in human cancer cells. Purity 99.8%
Catalog No. 50-287-9706 Supplier TARGETMOL CHEMICALS INC Supplier No. T71372MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. I-191 is a potent antagonist of protease-activated receptor 2 (PAR2), inhibits multiple signaling functions in human cancer cells. Purity 99.8%