Learn More
Medchemexpress LLC HY-117793 5mg Medchemexpress, I-191 CAS:1690172-25-8 Purity:>98%
SDP
Catalog No. 501932529
Encompass_Preferred
Medchemexpress, HY-117793 5mg I-191 CAS:1690172-25-8 I-191 is a potent, selective protease-activated receptor 2 (PAR2) antagonist [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2529 Supplier Medchemexpress LLC Supplier No. HY1177935MG
Medchemexpress, HY-117793 5mg I-191 CAS:1690172-25-8 I-191 is a potent, selective protease-activated receptor 2 (PAR2) antagonist [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.