Learn More
Selleck Chemical LLC I-191-E0102-100MG
SDP
Catalog No. 503135938
Encompass_Preferred
I-191 is a potent PAR2 antagonist that inhibits multiple PAR2-induced signaling pathways and functional responses I-191 also inhibits ERK1/2 phosphorylation RhoA activation and inhibition of forskolin-stimulated cAMP accumulation induced by micromolar concentrations of biased ligand GB88
Catalog No. 50-313-5938 Supplier Selleck Chemical LLC Supplier No. E0102100MG
I-191 is a potent PAR2 antagonist that inhibits multiple PAR2-induced signaling pathways and functional responses I-191 also inhibits ERK1/2 phosphorylation RhoA activation and inhibition of forskolin-stimulated cAMP accumulation induced by micromolar concentrations of biased ligand GB88