Learn More
Medchemexpress LLC I-287 | 2014368-93-3 | 99.4% | 565.04 | 50 MG
SDP
Catalog No. 5000361339
Encompass_Preferred
I-287 is an orally active, selective PAR2 inhibitor that functions as a negative allosteric regulator of Gαq and Gα12/13 activity and their downstream effectors. It has been shown to reduce Complete Freund's adjuvant (CFA)-induced inflammation in mice and is suitable for inflammation/immunology research.
  • Selectively targets Gαq and Gα12/13 signaling
  • Inhibits PAR2-mediated activation of proteins without impacting βarrestin2 recruitment or receptor internalization
  • Inhibits PAR2-mediated ERK1/2 activation in HEK293 cells
  • Inhibits PAR2-induced secretion of IL-8 cytokine in HCT-116 and A549 cells
  • Reduces inflammation in C57BL/6J mice
Catalog No. 50-003-61339 Supplier Medchemexpress LLC Supplier No. HY14801650MG
I-287 is an orally active, selective PAR2 inhibitor that functions as a negative allosteric regulator of Gαq and Gα12/13 activity and their downstream effectors. It has been shown to reduce Complete Freund's adjuvant (CFA)-induced inflammation in mice and is suitable for inflammation/immunology research.
  • Selectively targets Gαq and Gα12/13 signaling
  • Inhibits PAR2-mediated activation of proteins without impacting βarrestin2 recruitment or receptor internalization
  • Inhibits PAR2-mediated ERK1/2 activation in HEK293 cells
  • Inhibits PAR2-induced secretion of IL-8 cytokine in HCT-116 and A549 cells
  • Reduces inflammation in C57BL/6J mice