Learn More
Medchemexpress LLC HY-18407 100mg Medchemexpress, N-Methylisatoic anhydride CAS:10328-92-4 Purity:>98%
SDP
Catalog No. 501970008
Encompass_Preferred
Medchemexpress, HY-P7198 10ug I-309/CCL1 (CHO-expressed), Human CAS: Recombinant Human I-309/CCL1 (CHO-expressed) attracts monocytes, NK cells, and immature B cells and dendritic cells by interacting with a cell surface chemokine receptor called CCR8. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0008 Supplier Medchemexpress LLC Supplier No. HY18407100MG
Add to Cart
Add to Cart
Medchemexpress, HY-P7198 10ug I-309/CCL1 (CHO-expressed), Human CAS: Recombinant Human I-309/CCL1 (CHO-expressed) attracts monocytes, NK cells, and immature B cells and dendritic cells by interacting with a cell surface chemokine receptor called CCR8. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.