Learn More
Percival Scientific Percival Scientific I-36VL Environmental Chamber, IntellusUltra Controller, Temperature Range +2°C to 44°C, 29.7 cu ft
SDP
Catalog No. NC1969490
Encompass_Preferred
The PERCIVAL SCIENTIFIC I-36VL Environmental Chamber is a versatile refrigerated incubator designed for scientific applications such as bioassays and seed germination. It offers a controlled environment with robust construction and advanced temperature control, differentiating itself with features like programmable lighting and easy maintenance access. Its capacity of 29.7 cu ft and adjustable shelving enhance functionality, ensuring it meets diverse laboratory needs.
Catalog No. NC1969490 Supplier Percival Scientific Supplier No. I36VL
Add to Cart
Add to Cart
Request a Quote
The PERCIVAL SCIENTIFIC I-36VL Environmental Chamber is a versatile refrigerated incubator designed for scientific applications such as bioassays and seed germination. It offers a controlled environment with robust construction and advanced temperature control, differentiating itself with features like programmable lighting and easy maintenance access. Its capacity of 29.7 cu ft and adjustable shelving enhance functionality, ensuring it meets diverse laboratory needs.