Learn More
BUCHI I-80 Controller for Rotavapor R-80
SDP
Catalog No. 01672997 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Rotavapor R-80 House Vacuum
Rotavapor R-80 V-80/180
Includes:
Communication Cable, Holder
Woulff Bottle, Communication Cable, Tubing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Includes
01-672-997 Rotavapor R-80 House Vacuum Woulff Bottle, Communication Cable, Tubing
01-672-996 Rotavapor R-80 V-80/180 Communication Cable, Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-672-997 Supplier BUCHI Supplier No. 11I0800
Add to Cart
Add to Cart
The I-80 is an easy-to-use interface for conveniently regulating your vacuum manually. Increase control of the evaporation process by setting the pump speed and adjusting the manometer.
  • Functional design fulfills essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to centralized control of major process parameters
  • Full process control thanks to parameter monitoring
  • Proven technology for major applications
Specifications
Type Controller Interface
For Use With (Equipment) Rotavapor R-80 House Vacuum
Includes Woulff Bottle, Communication Cable, Tubing