Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Bacillus subtilis Z094, Genomic DNA
SDP

Catalog No. 22157506 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-506 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-506 Supplier ZeptoMetrix Supplier No. 0801562DNA10UG
Add to Cart
Add to Cart

For use as an amplification and/or detection control for nucleic acid testing of Bacillus subtilis

  • Can also be used to determine a limit of detection (LOD), in diagnostic assay development, cross-reactivity studies or genomic sequencing
  • Each aliquot contains 10μg of DNA extracted from pure culture
  • Supplied in TE Buffer

Controls should be run using the same protocols as those used to amplify extracted clinical specimens

Amplification/detection control for nucleic acid testing of Bacillus subtilis

Specifications

Certifications/Compliance ISO 9001:2008, ISO 13485:2003
Content And Storage -20°C
Form Solution
Product Type Genomic DNA Bacteria Control
For Use With (Application) Amplification/detection control for Nucleic Acid testing of Bacillus subtilis
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.