Learn More
Selleck Chemical LLC Molibresib (I-BET-762) 2mg 1260907-17-2 GSK525762, GSK525762A
SDP
Supplier:  Selleck Chemical LLC S71892MG
Encompass_Preferred
Molibresib (I-BET-762, GSK525762, GSK525762A) is an inhibitor for BET proteins with IC50 of ~35 nM in a cell-free assay, suppresses the production of proinflammatory proteins by macrophages and blocks acute inflammation, highly selective over other bromodomain-containing proteins. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7974