Learn More
Medchemexpress LLC I-BET151 | 1300031-49-5 | 99.47% | 415.44 | 100 MG
SDP
Catalog No. 5000364830
Encompass_Preferred
I-BET151 (GSK1210151A) is a BET bromodomain inhibitor that inhibits BRD4, BRD2, and BRD3. It is intended for research use only and has shown antiproliferative activity in various cell lines, as well as reduced tumor size in in vivo models.
  • Inhibits BRD4, BRD2, and BRD3
  • Shows antiproliferative activity in various cell lines
  • Significantly decreases cell proliferation and tumor size in in vivo models
  • Exhibits low blood clearance in rats and good oral systemic exposure
Catalog No. 50-003-64830 Supplier Medchemexpress LLC Supplier No. 100MGHY13235
I-BET151 (GSK1210151A) is a BET bromodomain inhibitor that inhibits BRD4, BRD2, and BRD3. It is intended for research use only and has shown antiproliferative activity in various cell lines, as well as reduced tumor size in in vivo models.
  • Inhibits BRD4, BRD2, and BRD3
  • Shows antiproliferative activity in various cell lines
  • Significantly decreases cell proliferation and tumor size in in vivo models
  • Exhibits low blood clearance in rats and good oral systemic exposure