Learn More
Selleck Chemical LLC I-BET151 (GSK1210151A) 5mg 1300031-49-5
SDP
Supplier:  Selleck Chemical LLC S27805MG
Encompass_Preferred
I-BET151 (GSK1210151A) is a novel selective BET inhibitor for BRD2, BRD3 and BRD4 with IC50 of 0.5 ?M, 0.25 ?M, and 0.79 ?M in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0954228