Learn More
Apexbio Technology LLC I-BET151 (GSK1210151A), 1300031-49-5, MFCD22124472, 10 mg
SDP
Supplier:  Apexbio Technology LLC B150010
Encompass_Preferred
MF: C23H21N5O3, MW: 415.44, Solubility: 41.5mg/mL in DMSO. I-BET151 (GSK1210151A) is a selective inhibitor of BET with pIC50 value of 6.1.
Catalog No. 50-101-4667