Learn More
Medchemexpress LLC I-bu-rG phosphoramidite | 147201-04-5 | 970.18 | 1 G
SDP
Catalog No. 5000446030
Encompass_Preferred
I-bu-rG phosphoramidite is a phosphinamide monomer used in the synthesis of nucleotides and nucleic acids. It is for research use only.
  • Used in the synthesis of nucleotides and nucleic acids
  • Targets DNA/RNA synthesis
  • Acts as a nucleoside antimetabolite/analog
  • Appears as a white to yellow solid
  • Has a purity of 98.0%
Catalog No. 50-004-46030 Supplier Medchemexpress LLC Supplier No. HYW0061031G
I-bu-rG phosphoramidite is a phosphinamide monomer used in the synthesis of nucleotides and nucleic acids. It is for research use only.
  • Used in the synthesis of nucleotides and nucleic acids
  • Targets DNA/RNA synthesis
  • Acts as a nucleoside antimetabolite/analog
  • Appears as a white to yellow solid
  • Has a purity of 98.0%