Learn More
Thermo Scientific™ Burkholderia cepacia Selective Agar Plated Medium (USP 60) 60mm
Shop All Thermo Scientific ProductsDescription
Burkholderia cepacia Selective Agar Plated Medium (USP 60) 60mm is recommended for the qualitative procedures for the selective and differential isolation of B. cepacia complex in purified water systems used in the manufacture of finished products for inhalation use of non-sterile aqueous preparations for oral, oromucosal, cutaneous or nasal use.
- Compliant formulation according to USP 60
- Quality tested to meet USP 60 specifically for Pharma customers
- Part of a complete workflow which include DCM and QCO
Specifications
Specifications
| Description | Burkholderia cepacia Sel Agar (USP60) (60mm) 10/PK |
| Format | 60 mm petri dish |
| Product Type | Agar Plate |
| For Use With (Application) | Pharmaceutical purified water testing used in non-sterile manufacturing. |
| Quantity | 10/Pk. |
| Regulatory Status | Lab use only |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.