Learn More
Waters Corp Waters Corporation I-Button, CH-A, Spare Parts Kit
Catalog No. 50793081 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This I-Button is a spare part designated as CH-A. It is categorized as a spare parts kit and is used with the ACQUITY UPLC H-Class System, specifically with the ACQUITY H-Class Column Heater Active.
  • Spare parts kit for maintenance and repair
  • Designated as CH-A
  • Compatible with ACQUITY UPLC H-Class System
  • Used with ACQUITY H-Class Column Heater Active
Catalog No. 50-793-081 Supplier Waters Corp Supplier No. 700005251
This I-Button is a spare part designated as CH-A. It is categorized as a spare parts kit and is used with the ACQUITY UPLC H-Class System, specifically with the ACQUITY H-Class Column Heater Active.
  • Spare parts kit for maintenance and repair
  • Designated as CH-A
  • Compatible with ACQUITY UPLC H-Class System
  • Used with ACQUITY H-Class Column Heater Active