Learn More
Description
- Each frozen aliquot contains 1μg of DNA extracted from pure culture of a recombinant strain of Saccharomyces cerevisiae
- Identification of the organism was confirmed by rDNA sequencing
- Used to determine limit of detection (LOD), in diagnostic assay development, cross-reactivity studies or genomic sequencing
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Specifications
Specifications
| Biosafety Level | 1 |
| Certifications/Compliance | ISO, 13485 |
| Content And Storage | -65°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | Titered Live Bacteria Control |
| For Use With (Application) | Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
