Learn More
Selleck Chemical LLC I-CBP112
SDP
Supplier:  Selleck Chemical LLC S67582MG
Encompass_Preferred
I-CBP112 is a potent and selective CBP/p300 inhibitor with dissociation constant (KD) of 151 6 nM and 167 8 nM for CBP and p300 respectively
Catalog No. 50-000-02897