Learn More
TARGETMOL CHEMICALS INC I-CBP112 HYDROCHLORIDE 5MG
SDP
Catalog No. 502579919
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. I-CBP112 is a selective inhibitor of the bromodomain-containing transcription factors. I-CBP112 (1 mM) has little activity against other bromodomains. I-CBP112 targets the CBP/p300 bromodomains. I-CBP112 significantly reduced the leukemia-initiating potential of MLL-AF9(+) acute myeloid leukemia cells in a dose-dependent manner in vitro and in vivo. Interestingly I-CBP112 increased the cytotoxic activity of BET bromodomain inhibitor JQ1 as well as doxorubicin. purity: 97%
Catalog No. 50-257-9919 Supplier TARGETMOL CHEMICALS INC Supplier No. T42475MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. I-CBP112 is a selective inhibitor of the bromodomain-containing transcription factors. I-CBP112 (1 mM) has little activity against other bromodomains. I-CBP112 targets the CBP/p300 bromodomains. I-CBP112 significantly reduced the leukemia-initiating potential of MLL-AF9(+) acute myeloid leukemia cells in a dose-dependent manner in vitro and in vivo. Interestingly I-CBP112 increased the cytotoxic activity of BET bromodomain inhibitor JQ1 as well as doxorubicin. purity: 97%