Learn More
New England Biolabs, Inc. I-CeuI – 500 units
SDP
Catalog No. 50812406
Encompass
A restriction endonuclease that recognizes the sequence CGTAACTATAACGGTC_CTAA^GGTAGCGAA.The intron encoding I-CeuI is present in the chloroplast large rRNA gene of Chlamydomonas eugametos (1). This gene has been cloned and overexpressed in E. coli (2).
Catalog No. 50-812-406 Supplier New England Biolabs, Inc. Supplier No. R0699S
A restriction endonuclease that recognizes the sequence CGTAACTATAACGGTC_CTAA^GGTAGCGAA.The intron encoding I-CeuI is present in the chloroplast large rRNA gene of Chlamydomonas eugametos (1). This gene has been cloned and overexpressed in E. coli (2).