Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Chlamydia trachomatis IgM ELISA Kit

Catalog No. 89032281 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-281 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-281 Supplier Abnova™ Supplier No. KA4827
Only null left
Add to Cart
Add to Cart

Enzyme immunoassay for the detection of IgM antibodies to Chlamydia trachomatis in human serum or plasma

Intended for detection of specific IgM antibodies in a sample by means of a sandwich type of the EIA method (i.e. a solid phase coated with specific antigen – antibody from the analyzed sample – labeled antibody).

Specifications

Description Chlamydia trachomatis IgM ELISA Kit is an enzyme-linked immunosorbent assay for the detection of IgM antibodies to Chlamydia trachomatis.
Detection Method Colorimetric
Product Type Chlamydia trachomatis IgM ELISA Kit
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) Qualitative
Gene ID (Entrez) Microtitre Plate: 96 wells, Negative Control: (2mL), CUT-OFF: (3mL), Positive Control: (2mL), Conjugate: 15mL, Sample Diluent 2: 105mL, TMB-Complete 2: 15mL, Wash Solution: 20x concentrated buffer: 75mL, Stop Solution: Acid solution: 15mL
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store the kit at 4°C. Do not freeze.
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.