Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Escherichia coli O111:NM, Genomic DNA
SDP

Catalog No. 22157900 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-900 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Not intended for use in manufacture or processing of injectable products subject to licensure under section 351 of Public Health Service Act or for any other product intended for administration to humans.
Catalog No. 22-157-900 Supplier ZeptoMetrix Supplier No. 0801747DNA10UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Not intended for use in manufacture or processing of injectable products subject to licensure under section 351 of Public Health Service Act or for any other product intended for administration to humans.

Use as an amplification or detection control for nucleic acid testing of Escherichia coli

  • Extracted from pure culture of Escherichia coli.
  • Also used to determine limit of detection (LOD) in diagnostic assay development, cross-reactivity studies or genomic sequencing

Specifications

Certifications/Compliance OSHA 1910.1200
Format Liquid
pH 7 to 8
Product Type Genomic DNA
Source Escherichia coli O111:NM
Storage Buffer TE Buffer
Content And Storage -20°C
Quantity 10 μg
For Use With (Application) Amplification/detection control for Nucleic Acid testing of E. coli., determines limit of detection in diagnostic assay, cross-reactivity studies or genomic sequencing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.