Learn More
Med Vet International I-Explore Microscope
Catalog No. 503046147
Encompass_Preferred
I-EXPLORE (Model SMD-04) is designed for viewing solid or three dimensional objects rather than glass slides. This scope makes it possible to view virtually anything magnified 20X. The binocular head allows comfortable viewing with both eyes (no more squinting) and the built in light illuminates all specimens for premium viewing every time. Real glass optics provide high quality images and battery power makes it portable for outdoor use.Microscope package includes:Binocular eyetubesBuilt in light sourceReal optical glass lenses20X magnificationPortable battery powerCE CertifiedCPSIA approved
Catalog No. 50-304-6147 Supplier Med Vet International Supplier No. SMD04
I-EXPLORE (Model SMD-04) is designed for viewing solid or three dimensional objects rather than glass slides. This scope makes it possible to view virtually anything magnified 20X. The binocular head allows comfortable viewing with both eyes (no more squinting) and the built in light illuminates all specimens for premium viewing every time. Real glass optics provide high quality images and battery power makes it portable for outdoor use.Microscope package includes:Binocular eyetubesBuilt in light sourceReal optical glass lenses20X magnificationPortable battery powerCE CertifiedCPSIA approved