Learn More
Pyramex™ I-Force™ Slim Safety Eyewear
Catalog No. 19155810 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Indoor/Outdoor Mirror, Mirror
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-810 Indoor/Outdoor Mirror, Mirror
19-155-805 Clear
19-155-807 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-810 Supplier Pyramex™ Supplier No. SB7080SDNT
Add to Cart
Add to Cart
This item is not returnable. View return policy
I-Force™ Slim Safety Eyewear with sporty, lightweight design eliminates the bulkiness and weight of a goggle.
  • Designed for slimmer faces, the I-Force™ Slim is about 6% smaller than the I-Force™
  • Ratcheting Quick Release interchangeable temples and strap
  • Vented foam carriage
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • H2X anti-fog lenses
  • Dual pane lens provides the ultimate anti-fog performance
  • Outer polycarbonate lens protects against the environment, while inner acetate lens is designed to prevent fogging
  • Outside lens is coated with H2X Anti-fog Technology
  • Passes MIL-PRF 32432 High Velocity Impact Standards
Specifications
Certifications/Compliance ANSI Z87.1, CSA Z94.3-07
Lens Coating Anti-fog, UV-treated, Scratch-resistant, Impact-resistant
Certifications/Compliance (Europe) EN-166
Lens Tint Indoor/Outdoor Mirror, Mirror
Product Type Ballistic Safety Glasses
Frame Color Black
Material Polycarbonate, Nylon
Disposable Reusable
Lens Material Polycarbonate (Front Lens), Acetate (Inner Lens), PU Foam
Frame Material Nylon
UV Protection U6
Frame Type Full Frame
Autoclavable Not Autoclavable