Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Gardnerella vaginalis Z125, Titered
SDP

Catalog No. 22157901 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-901 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Not intended for use in manufacture or processing of injectable products subject to licensure under section 351 of Public Health Service Act or for any other product intended for administration to humans. Must be used within Biological Safety Level 2 facility or cabinet.
Catalog No. 22-157-901 Supplier ZeptoMetrix Supplier No. 0801894
Add to Cart
Add to Cart
This item is not returnable. View return policy
Not intended for use in manufacture or processing of injectable products subject to licensure under section 351 of Public Health Service Act or for any other product intended for administration to humans. Must be used within Biological Safety Level 2 facility or cabinet.

Used for determination of limit of detection (LOD) in diagnostic assays or crossreactivity studies

  • Biosafety level 2 microorganism
  • Available in 1mL aliquot
  • 16S sequencing used to confirm identification of organism
  • Gram staining and additional culturing monitors purity of culture
  • Glycerol (15%) used as cryoprotectant for freezing medium
  • Also used as control for nucleic acid tests

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Content And Storage -65°C
Form Solution
Product Type Titered Live Bacteria Control
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.